GBT-440 98%

Supplier: Ambeed
Warning

Synonyms: Benzaldehyde, Voxelotor, Hemoglobin Modulators-1, 2-Hydroxy-6-((2-(1-(1-methylethyl)-1H-pyrazol-5-yl)-3-pyridinyl)methoxy)-, 2-hydroxy-6-((2-(1-isopropyl-1H-pyrazol-5-yl)pyridin-3-yl)methoxy)benzaldehyde, 2-hydroxy-6-((2-(1-(propan-2-yl)-1H-pyrazol-5-yl)pyridin-3-yl)methoxy)benzaldehyde, 2-Hydroxy-6-[[2-(2-propan-2-ylpyrazol-3-yl)pyridin-3-yl]methoxy]benzaldehyde

A222549-10MG A222549-25MG A222549-5MG A222549-100MG A222549-1G A222549-1MG A222549-250MG
77237-652EA 152.44 USD
77237-652 77237-654 77237-656 77746-313 77746-314 77746-315 77746-316
GBT-440 98%
GBT-440

Precaución: for Research Use Only

Formula: C₁₉H₁₉N₃O₃
MW: 337.37 g/mol
MDL Number: MFCD29053764
CAS Number: 1446321-46-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR