p-Toluenesulfonylmethyl chloride 98%

Supplier: Ambeed
Warning

Synonyms: 1-((Chloromethyl)sulphonyl)-4-methylbenzene, Chloromethyl p-Tolyl Sulphone, p-Tolylsulphonylmethyl Chloride

A222545-1G A222545-5G
77249-416EA 6.18 USD
77249-416 77568-150
p-Toluenesulfonylmethyl chloride 98%
p-Toluenesulfonylmethyl chloride

Precaución: For research use only.

Formula: C₈H₉ClO₂S
MW: 204.67 g/mol
MDL Number: MFCD00134281
CAS Number: 7569-26-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR