5-Hydroxynicotinic acid 97%

Supplier: Ambeed
Warning

Synonyms: 5-Hydroxypyridine-3-carboxylic acid, 5-Hydroxy-3-pyridinecarboxylic acid

A222460-10G A222460-25G A222460-500G A222460-5G
77275-500EA 10.3 USD
77275-500 77275-502 77275-504 77275-506
5-Hydroxynicotinic acid 97%
5-Hydroxynicotinic acid

Precaución: for Research Use Only

Formula: C₆H₅NO₃
MW: 139.11 g/mol
MDL Number: MFCD00129117
CAS Number: 27828-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR