4-(Methylthio)phenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-(Methylthio)benzeneboronic acid, 4-Thioanisoleboronic acid

A222152-100G A222152-10G A222152-1G A222152-25G A222152-5G
77275-490EA 204.97 USD
77275-490 77275-492 77275-494 77275-496 77275-498
4-(Methylthio)phenylboronic acid 97%
4-(Methylthio)phenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₉BO₂S
MW: 168.02 g/mol
MDL Number: MFCD00093410
CAS Number: 98546-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR