4-Bromo-4'-nitrobiphenyl 95%

Supplier: Ambeed
Warning

A221655-100MG A221655-250MG
77288-234EA 20.6 USD
77288-234 77288-236
4-Bromo-4'-nitrobiphenyl 95%
4-Bromo-4'-nitrobiphenyl

Precaución: for Research Use Only

Formula: C₁₂H₈BrNO₂
MW: 278.1014 g/mol
MDL Number: MFCD00024697
CAS Number: 6242-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR