Methanesulfonato[9,9-dimethyl-4,5-bis(diphenylphosphino)xanthene](2'-methylamino-1,1'-biphenyl-2-yl)palladium(II) 98%

Supplier: Ambeed
Warning

Synonyms: XantPhos Pd G4

A221417-100MG A221417-1G A221417-250MG A221417-5G
77249-394EA 30.9 USD
77249-394 77249-396 77249-398 77249-400
Methanesulfonato[9,9-dimethyl-4,5-bis(diphenylphosphino)xanthene](2'-methylamino-1,1'-biphenyl-2-yl)palladium(II) 98%
Methanesulfonato[9,9-dimethyl-4,5-bis(diphenylphosphino)xanthene](2'-methylamino-1,1'-biphenyl-2-yl)palladium(II)

Precaución: for Research Use Only

Formula: C₅₃H₄₇NO₄P₂PdS
MW: 962.38 g/mol
MDL Number: MFCD28411658
CAS Number: 1621274-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR