5-Bromo-6-chloro-2,3-dihydro-1H-inden-1-one 97%

Supplier: Ambeed
Warning

A221364-100MG A221364-1G A221364-250MG A221364-5G
77249-388EA 43.26 USD
77249-388 77249-390 77249-392 77568-134
5-Bromo-6-chloro-2,3-dihydro-1H-inden-1-one 97%
5-Bromo-6-chloro-2,3-dihydro-1H-inden-1-one

Precaución: For research use only.

Formula: C₉H₆BrClO
MW: 245.50 g/mol
MDL Number: MFCD18208169
CAS Number: 1260018-10-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR