Methyl-3,4-dihydroxybenzoate 98%

Supplier: Ambeed
Warning

Synonyms: 3,4-Dihydroxybenzoic acid methyl ester, Methyl 3,4-dihydroxybenzoate

A221114-100G A221114-1G A221114-25G A221114-500G A221114-50MG A221114-5G
77249-374EA 93.73 USD
77249-374 77249-376 77249-378 77249-380 77249-382 77249-384
Methyl-3,4-dihydroxybenzoate 98%
Methyl-3,4-dihydroxybenzoate

Precaución: for Research Use Only

Formula: C₈H₈O₄
MW: 168.15 g/mol
MDL Number: MFCD00016437
CAS Number: 2150-43-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR