2-Amino-1-(3-((4-chlorophenyl)amino)-2-(4-fluorophenyl)-8,8-dimethyl-5,6-dihydroimidazo[1,2-a]pyrazin-7(8H)-yl)ethanone ≥98%
Supplier: Ambeed
|
Warning
|
A220397-10MG
A220397-1MG
A220397-25MG
77237-626EA
299
USD
77237-626
77237-628
77237-630
77237-632
2-Amino-1-(3-((4-chlorophenyl)amino)-2-(4-fluorophenyl)-8,8-dimethyl-5,6-dihydroimidazo[1,2-a]pyrazin-7(8H)-yl)ethanone ≥98%
2-Amino-1-(3-((4-chlorophenyl)amino)-2-(4-fluorophenyl)-8,8-dimethyl-5,6-dihydroimidazo[1,2-a]pyrazin-7(8H)-yl)ethanone
Precaución: for Research Use Only
Formula:
C₂₂H₂₃ClFN₅O |
MDL Number:
MFCD28167758 CAS Number: 1261114-01-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...