4-Chloro-3-fluoroanisole 98%

Supplier: Ambeed
Warning

Synonyms: 1-Chloro-2-fluoro-4-methoxybenzene

A220176-100G A220176-10G A220176-1G A220176-25G A220176-5G
77288-202EA 230.72 USD
77288-202 77288-204 77288-206 77288-208 77288-210
4-Chloro-3-fluoroanisole 98%
4-Chloro-3-fluoroanisole

Precaución: for Research Use Only

Formula: C₇H₆ClFO
MW: 160.58 g/mol
MDL Number: MFCD00042570
CAS Number: 501-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR