1-(4-Amino-3-chloro-5-(trifluoromethyl)phenyl)ethanone 97%

Supplier: Ambeed
Warning

A220118-100MG A220118-1G A220118-250MG A220118-5G
77288-190EA 17.51 USD
77288-190 77288-192 77288-194 77288-196
1-(4-Amino-3-chloro-5-(trifluoromethyl)phenyl)ethanone 97%
1-(4-Amino-3-chloro-5-(trifluoromethyl)phenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₇ClF₃NO
MW: 237.61 g/mol
MDL Number: MFCD09907719
CAS Number: 97760-76-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR