(S)-(+)-2-Phenylglycine methyl ester hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: (S)-Methyl 2-amino-2-phenylacetate hydrochloride, H-Phg-OMe·HCl, L-(+)-2-Phenylglycine methyl ester hydrochloride

A219969-100G A219969-25G A219969-5G
77249-364EA 47.38 USD
77249-364 77249-366 77249-368
(S)-(+)-2-Phenylglycine methyl ester hydrochloride 97%
(S)-(+)-2-Phenylglycine methyl ester hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₂ClNO₂
MW: 201.65 g/mol
MDL Number: MFCD00077158
CAS Number: 15028-39-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR