3-(4-tert-Butylphenyl)isobutyraldehyde 98%

Supplier: Ambeed
Warning

A219864-25G A219864-1000G A219864-100G A219864-500G
77275-450EA 22 USD
77275-450 77615-840 77615-842 77615-844
3-(4-tert-Butylphenyl)isobutyraldehyde 98%
3-(4-tert-Butylphenyl)isobutyraldehyde

Precaución: for Research Use Only

Formula: C₁₂H₂₀O
MDL Number: MFCD00047655
CAS Number: 80-54-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR