1,8-Naphthyridine-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A219804-100MG A219804-1G A219804-250MG A219804-5G
77261-296EA 24 USD
77261-296 77261-298 77261-300 77261-302
1,8-Naphthyridine-2-carboxylic acid 97%
1,8-Naphthyridine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₆N₂O₂
MDL Number: MFCD00202900
CAS Number: 215523-34-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR