4-(1-Pyrrolidinyl)piperidine dihydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: 4-(Pyrrolidin-1-yl)piperidine dihydrochloride

A219766-10G A219766-1G A219766-250MG A219766-5G
77288-176EA 155 USD
77288-176 77288-178 77288-180 77288-182
4-(1-Pyrrolidinyl)piperidine dihydrochloride 95%
4-(1-Pyrrolidinyl)piperidine dihydrochloride

Precaución: for Research Use Only

Formula: C₉H₂₀Cl₂N₂
MW: 227.18 g/mol
MDL Number: MFCD00035294
CAS Number: 4983-39-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR