tert-Butyl ((2R,3R)-4-(4-acetamido-N-isobutylphenylsulfonamido)-3-hydroxy-1-phenylbutan-2-yl)carbamate 99%
Supplier: Ambeed
|
Warning
|
A219615-100MG
A219615-10G
A219615-1G
A219615-250MG
A219615-5G
77261-284EA
16
USD
77261-284
77261-286
77261-288
77261-290
77261-292
77261-294
tert-Butyl ((2R,3R)-4-(4-acetamido-N-isobutylphenylsulfonamido)-3-hydroxy-1-phenylbutan-2-yl)carbamate 99%
tert-Butyl ((2R,3R)-4-(4-acetamido-N-isobutylphenylsulfonamido)-3-hydroxy-1-phenylbutan-2-yl)carbamate
Precaución: for Research Use Only
Formula:
C₂₇H₃₉N₃O₆S |
MDL Number:
MFCD31381099 CAS Number: 2126144-72-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...