6-((2-((4-(2,4-Dichlorophenyl)-5-(5-methyl-1H-imidazol-2-yl)pyrimidin-2-yl)amino)ethyl)amino)nicotinonitrile monohydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: Laduviglusib monohydrochloride

A219562-10MG A219562-1MG A219562-2MG A219562-50MG A219562-5MG
77261-274EA 25.75 USD
77261-274 77261-276 77261-278 77261-280 77261-282
6-((2-((4-(2,4-Dichlorophenyl)-5-(5-methyl-1H-imidazol-2-yl)pyrimidin-2-yl)amino)ethyl)amino)nicotinonitrile monohydrochloride 98%
6-((2-((4-(2,4-Dichlorophenyl)-5-(5-methyl-1H-imidazol-2-yl)pyrimidin-2-yl)amino)ethyl)amino)nicotinonitrile monohydrochloride

Precaución: for Research Use Only

Formula: C₂₂H₁₉Cl₃N₈
MW: 501.79 g/mol
MDL Number: MFCD30489757
CAS Number: 1797989-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR