5-Iodo-1H-indole-3-carbaldehyde 97%

Supplier: Ambeed
Warning

A219415-1G A219415-250MG
77249-348EA 119 USD
77249-348 77249-350
5-Iodo-1H-indole-3-carbaldehyde 97%
5-Iodo-1H-indole-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₉H₆INO
MDL Number: MFCD15176154
CAS Number: 114144-17-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR