(E)-1-Chloro-2-(2-nitrovinyl)benzene 98%

Supplier: Ambeed
Warning

A219371-1G A219371-5G
77261-270EA 8.24 USD
77261-270 77261-272
(E)-1-Chloro-2-(2-nitrovinyl)benzene 98%
(E)-1-Chloro-2-(2-nitrovinyl)benzene

Precaución: for Research Use Only

Formula: C₈H₆ClNO₂
MW: 183.59 g/mol
MDL Number: MFCD00024820
CAS Number: 22568-07-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR