2-Bromo-5-methoxyphenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-5-methoxybenzeneboronic acid, 2-Bromo-5-methoxybenzene boronic acid

A218827-100MG A218827-10G A218827-1G A218827-250MG A218827-5G
77261-254EA 8.24 USD
77261-254 77261-256 77261-258 77261-260 77261-262
2-Bromo-5-methoxyphenylboronic acid 97%
2-Bromo-5-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BBrO₃
MW: 230.85 g/mol
MDL Number: MFCD06659859
CAS Number: 89694-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR