2,4-Dichloro-8-methoxyquinazoline 97%

Supplier: Ambeed
Warning

A218658-100MG A218658-250MG A218658-1G
77249-328EA 37.08 USD
77249-328 77249-330 77746-308
2,4-Dichloro-8-methoxyquinazoline 97%
2,4-Dichloro-8-methoxyquinazoline

Precaución: for Research Use Only

Formula: C₉H₆Cl₂N₂O
MW: 229.0600 g/mol
MDL Number: MFCD09954882
CAS Number: 61948-60-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR