4-Bromo-1-methyl-1H-pyrazole-5-carboxylic acid 95%

Supplier: Ambeed
Warning

A218585-1G A218585-250MG A218585-5G
77288-168EA 35.02 USD
77288-168 77288-170 77748-348
4-Bromo-1-methyl-1H-pyrazole-5-carboxylic acid 95%
4-Bromo-1-methyl-1H-pyrazole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₅BrN₂O₂
MDL Number: MFCD00463982
CAS Number: 84547-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR