tert-Butyl 6-chloropyridazine-3-carboxylate 97%

Supplier: Ambeed
Warning

A218504-1G A218504-250MG
77237-602EA 280.16 USD
77237-602 77237-604
tert-Butyl 6-chloropyridazine-3-carboxylate 97%
tert-Butyl 6-chloropyridazine-3-carboxylate

Precaución: for Research Use Only

Formula: C₉H₁₁ClN₂O₂
MW: 214.64 g/mol
MDL Number: MFCD18897601
CAS Number: 1340506-55-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR