2,6-Dichloro-3-iodopyridine 98%

Supplier: Ambeed
Warning

A218472-100G A218472-10G A218472-1G A218472-250MG A218472-25G A218472-5G
77288-156EA 238.96 USD
77288-156 77288-158 77288-160 77288-162 77288-164 77288-166
2,6-Dichloro-3-iodopyridine 98%
2,6-Dichloro-3-iodopyridine

Precaución: for Research Use Only

Formula: C₅H₂Cl₂IN
MW: 273.88 g/mol
MDL Number: MFCD03094686
CAS Number: 148493-37-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR