Ethyl-2-aminopyrimidine-5-carboxylate 97%

Supplier: Ambeed
Warning

A218468-10G A218468-5G
77261-252EA 193 USD
77261-252 77593-422
Ethyl-2-aminopyrimidine-5-carboxylate 97%
Ethyl-2-aminopyrimidine-5-carboxylate

Precaución: for Research Use Only

Formula: C₇H₉N₃O₂
MDL Number: MFCD04114100
CAS Number: 57401-76-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR