3-Chloro-5-nitro-1H-pyrazole 97%

Supplier: Ambeed
Warning

A218388-100MG A218388-250MG A218388-1G
77237-598EA 27 USD
77237-598 77237-600 77746-303
3-Chloro-5-nitro-1H-pyrazole 97%
3-Chloro-5-nitro-1H-pyrazole

Precaución: for Research Use Only

Formula: C₃H₂ClN₃O₂
MDL Number: MFCD29762425
CAS Number: 1369959-12-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR