2,6-Dichloro-3-fluoropyridine 96%

Supplier: Ambeed
Danger

A218284-10G A218284-1G A218284-250MG A218284-25G A218284-5G A218284-100G
77249-316EA 39 USD
77249-316 77249-318 77249-320 77249-322 77249-324 77615-836
2,6-Dichloro-3-fluoropyridine 96%
2,6-Dichloro-3-fluoropyridine

Precaución: for Research Use Only

Formula: C₅H₂Cl₂FN
MDL Number: MFCD09907815
CAS Number: 52208-50-1
UN: 2923
ADR: 8,6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR