3-Bromo-5-chloro-1H-indazole 95%

Supplier: Ambeed
Warning

A218157-1G A218157-250MG A218157-5G
77275-412EA 70.04 USD
77275-412 77275-414 77275-416
3-Bromo-5-chloro-1H-indazole 95%
3-Bromo-5-chloro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₄BrClN₂
MDL Number: MFCD07781584
CAS Number: 885521-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR