4,4',4''-(1,3,5-Triazine-2,4,6-triyl)trianiline 95%

Supplier: Ambeed
Warning

A218110-10G A218110-1G A218110-250MG A218110-25G A218110-5G
77249-306EA 268.83 USD
77249-306 77249-308 77249-310 77249-312 77249-314
4,4',4''-(1,3,5-Triazine-2,4,6-triyl)trianiline 95%
4,4',4''-(1,3,5-Triazine-2,4,6-triyl)trianiline

Precaución: for Research Use Only

Formula: C₂₁H₁₈N₆
MW: 354.42 g/mol
MDL Number: MFCD04116311
CAS Number: 14544-47-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR