2-Chloro-5-(trifluoromethyl)pyridine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-5-trifluoromethylpyridine

A218085-1000G A218085-100G A218085-10G A218085-25G A218085-500G
77275-402EA 157.59 USD
77275-402 77275-404 77275-406 77275-408 77275-410
2-Chloro-5-(trifluoromethyl)pyridine 98%
2-Chloro-5-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₃ClF₃N
MW: 181.54 g/mol
MDL Number: MFCD00042225
CAS Number: 52334-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR