Tris(4-methoxyphenyl)amine 97%

Supplier: Ambeed
Warning

A218053-250MG A218053-1G
77249-304EA 128 USD
77249-304 77550-408
Tris(4-methoxyphenyl)amine 97%
Tris(4-methoxyphenyl)amine

Precaución: for Research Use Only

Formula: C₂₁H₂₁NO₃
MW: 335.39 g/mol
CAS Number: 13050-56-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR