(S)-2,2'-Bis[bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-4,4',6,6'-tetramethoxybiphenyl 98%

Supplier: Ambeed
Warning

Synonyms: (S)-(4,4',6,6'-Tetramethoxybiphenyl-2,2'-diyl)bis[bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphine], (S)-DTBM-Garphos

A217855-100MG A217855-1G A217855-250MG
77249-298EA 90 USD
77249-298 77249-300 77249-302
(S)-2,2'-Bis[bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-4,4',6,6'-tetramethoxybiphenyl 98%
(S)-2,2'-Bis[bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-4,4',6,6'-tetramethoxybiphenyl

Precaución: for Research Use Only

Formula: C₇₆H₁₀₈O₈P₂
MW: 1211.61 g/mol
MDL Number: MFCD20265636
CAS Number: 1365531-99-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR