2-Bromo-3,4-dimethoxybenzaldehyde 95%

Supplier: Ambeed
Warning

A217802-250MG A217802-1G
77261-244EA 37 USD
77261-244 77550-404
2-Bromo-3,4-dimethoxybenzaldehyde 95%
2-Bromo-3,4-dimethoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉BrO₃
MDL Number: MFCD07772896
CAS Number: 55171-60-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR