3-Benzyloxy-4-methylphenylamine 95%

Supplier: Ambeed
Warning

A217797-1G A217797-250MG A217797-5G
77275-380EA 80 USD
77275-380 77275-382 77275-384
3-Benzyloxy-4-methylphenylamine 95%
3-Benzyloxy-4-methylphenylamine

Precaución: for Research Use Only

Formula: C₁₄H₁₅NO
MDL Number: MFCD04973042
CAS Number: 219492-12-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR