3-Isopropylbenzene-1,2-diol 97%

Supplier: Ambeed
Warning

A217742-250MG A217742-5G
77275-376EA 75.99 USD
77275-376 77275-378
3-Isopropylbenzene-1,2-diol 97%
3-Isopropylbenzene-1,2-diol

Precaución: for Research Use Only

Formula: C₉H₁₂O₂
MDL Number: MFCD00016434
CAS Number: 2138-48-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR