2,7-Dimethylnaphthalene 97%

Supplier: Ambeed
Danger

A217739-10G A217739-1G A217739-250MG A217739-25G A217739-5G
77249-284EA 264.01 USD
77249-284 77249-286 77249-288 77249-290 77249-292
2,7-Dimethylnaphthalene 97%
2,7-Dimethylnaphthalene

Precaución: for Research Use Only

Formula: C₁₂H₁₂
MDL Number: MFCD00004121
CAS Number: 582-16-1
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR