3,5,5,8,8-Pentamethyl-5,6,7,8-tetrahydro-2-naphthaleneboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: (3,5,5,8,8-Pentamethyl-5,6,7,8-tetrahydronaphthalen-2-yl)boronic acid

A217668-250MG A217668-5G
77275-372EA 62 USD
77275-372 77275-374
3,5,5,8,8-Pentamethyl-5,6,7,8-tetrahydro-2-naphthaleneboronic acid 95%
3,5,5,8,8-Pentamethyl-5,6,7,8-tetrahydro-2-naphthaleneboronic acid

Precaución: for Research Use Only

Formula: C₁₅H₂₃BO₂
MW: 246.1529 g/mol
MDL Number: MFCD08436043
CAS Number: 169126-64-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR