2-Fluoro-5-nitrophenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-5-nitrobenzeneboronic acid

A217659-10G A217659-1G A217659-250MG A217659-25G A217659-5G
77275-362EA 39 USD
77275-362 77275-364 77275-366 77275-368 77275-370
2-Fluoro-5-nitrophenylboronic acid 98%
2-Fluoro-5-nitrophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₅BFNO₄
MDL Number: MFCD03092929
CAS Number: 819849-20-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR