2-Cyano-3-pyridinecarboxylic acid methyl ester 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-cyanonicotinate

A217628-100MG A217628-1G A217628-250MG
77261-232EA 40.17 USD
77261-232 77261-234 77261-236
2-Cyano-3-pyridinecarboxylic acid methyl ester 98%
2-Cyano-3-pyridinecarboxylic acid methyl ester

Precaución: for Research Use Only

Formula: C₈H₆N₂O₂
MW: 162.145 g/mol
MDL Number: MFCD00128863
CAS Number: 75358-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR