(±)-2,3-Dibromo-1-propanol 95%

Supplier: Ambeed
Danger

Synonyms: 2,3-Dibromo-1-propanol, Glycerol alpha,beta-Dibromohydrin, alpha,beta-Dibromohydrin

A217507-10G A217507-1G A217507-25G
77249-264EA 7.21 USD
77249-264 77249-266 77249-268
(±)-2,3-Dibromo-1-propanol 95%
(±)-2,3-Dibromo-1-propanol

Precaución: for Research Use Only

Formula: HOCH₂BrCHCH₂Br
MW: 217.89 g/mol
MDL Number: MFCD00004699
CAS Number: 96-13-9
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR