Perylene-3,9-dicarboxylic acid ≥95%

Supplier: Ambeed
Warning

A217468-100MG A217468-250MG
77249-260EA 14 USD
77249-260 77249-262
Perylene-3,9-dicarboxylic acid ≥95%
Perylene-3,9-dicarboxylic acid

Precaución: for Research Use Only

Formula: C₂₂H₁₂O₄
MW: 340.32 g/mol
MDL Number: MFCD00082168
CAS Number: 6364-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR