Cyclohexanone-4-carboxaldehyde 95%

Supplier: Ambeed
Warning

Synonyms: 4-Oxocyclohexanecarbaldehyde

A217417-100MG A217417-250MG A217417-5G
77288-106EA 31 USD
77288-106 77288-108 77288-110
Cyclohexanone-4-carboxaldehyde 95%
Cyclohexanone-4-carboxaldehyde

Precaución: for Research Use Only

Formula: C₇H₁₀O₂
MDL Number: MFCD04972553
CAS Number: 96184-81-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR