7-Methoxybenzofuran-3(2H)-one 95%

Supplier: Ambeed
Warning

A217348-100MG A217348-1G A217348-250MG A217348-5G
77261-216EA 15 USD
77261-216 77261-218 77261-220 77261-222
7-Methoxybenzofuran-3(2H)-one 95%
7-Methoxybenzofuran-3(2H)-one

Precaución: for Research Use Only

Formula: C₉H₈O₃
MDL Number: MFCD00274296
CAS Number: 7169-37-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR