5-Chloro-3-pyridineboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 5-Chloropyridine-3-boronic acid Pinacol ester, 2-(5-Chloro-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

A217124-10G A217124-1G A217124-250MG A217124-5G
77275-350EA 200 USD
77275-350 77275-352 77275-354 77748-342
5-Chloro-3-pyridineboronic acid pinacol ester 98%
5-Chloro-3-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₁H₁₅BClNO₂
MDL Number: MFCD06798284
CAS Number: 865186-94-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR