3-Cyanophenylacetic acid 98%
Supplier: Ambeed
|
Warning
|
A217046-1G
A217046-250MG
A217046-5G
77275-332EA
26
USD
77275-332
77275-334
77275-336
3-Cyanophenylacetic acid 98%
3-Cyanophenylacetic acid
Precaución: for Research Use Only
Formula:
C₉H₇NO₂ |
MDL Number:
MFCD06411079 CAS Number: 1878-71-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...