2-Amino-5-bromobenzoylpyridine 98%

Supplier: Ambeed
Warning

A217026-10G A217026-1G A217026-25G
77237-562EA 25 USD
77237-562 77237-564 77568-100
2-Amino-5-bromobenzoylpyridine 98%
2-Amino-5-bromobenzoylpyridine

Precaución: For research use only.

Formula: C₁₂H₉BrN₂O
MDL Number: MFCD07780283
CAS Number: 1563-56-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR