3-(2-Bromophenyl)pyrrolidine 97%

Supplier: Ambeed
Warning

A216929-1G A216929-250MG
77249-254EA 572.68 USD
77249-254 77249-256
3-(2-Bromophenyl)pyrrolidine 97%
3-(2-Bromophenyl)pyrrolidine

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrN
MW: 226.11 g/mol
MDL Number: MFCD06659031
CAS Number: 1203686-38-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR