(1S)-5,5',6,6',7,7',8,8'-Octahydro-3,3'-di-9-phenanthrenyl-[1,1'-binaphthalene]-2,2'-diol 98%
Supplier: Ambeed
|
Warning
|
A216553-100MG
A216553-1G
A216553-250MG
77249-246EA
82.4
USD
77249-246
77249-248
77249-250
(1S)-5,5',6,6',7,7',8,8'-Octahydro-3,3'-di-9-phenanthrenyl-[1,1'-binaphthalene]-2,2'-diol 98%
(1S)-5,5',6,6',7,7',8,8'-Octahydro-3,3'-di-9-phenanthrenyl-[1,1'-binaphthalene]-2,2'-diol
Precaución: for Research Use Only
Formula:
C₄₈H₃₈O₂ MW: 646.80 g/mol |
CAS Number:
1028416-48-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...