TMP-195 98%

Supplier: Ambeed
Warning

Synonyms: TMP195, N-(2-methyl-2-(2-phenyloxazol-4-yl)propyl)-3-(5-(trifluoromethyl)-1,2,4-oxadiazol-3-yl)benzamide

A216033-10MG A216033-1MG A216033-50MG A216033-5MG A216033-100MG A216033-250MG
77261-194EA 41.2 USD
77261-194 77261-196 77261-198 77261-200 77748-340 77748-341
TMP-195 98%
TMP-195

Precaución: for Research Use Only

Formula: C₂₃H₁₉F₃N₄O₃
MW: 456.42 g/mol
MDL Number: MFCD26522024
CAS Number: 1314891-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR