4-Fluoroisatin 95%

Supplier: Ambeed
Warning

Synonyms: 4-Fluoroindoline-2,3-dione

A215922-1G A215922-5G
77288-096EA 133.9 USD
77288-096 77288-098
4-Fluoroisatin 95%
4-Fluoroisatin

Precaución: for Research Use Only

Formula: C₈H₄FNO₂
MW: 165.1200 g/mol
MDL Number: MFCD01175824
CAS Number: 346-34-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR